Assay Details
Gene Symbol: Csnk1g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023102.2 | NP_075590.2 | 4 - 5 | 759 | 72 | |
GenBank mRNA | U22297.1 | - | 3 - 4 | 493 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
