Assay Details
Gene Symbol: Prkaa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023991.1 | NP_076481.1 | 7 - 8 | 1297 | 106 | |
XM_008763901.1 | XP_008762123.1 | 6 - 7 | 3026 | 106 | ||
GenBank mRNA | U12149.1 | - | 7 - 8 | 1482 | 106 | |
Z29486.1 | - | 7 - 8 | 1297 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
