Assay Details
Gene Symbol: Prkaa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023991.1 | NP_076481.1 | 8 - 9 | 1431 | 87 | |
XM_008763901.1 | XP_008762123.1 | 7 - 8 | 3160 | 87 | ||
GenBank mRNA | Z29486.1 | - | 8 - 9 | 1431 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
fatty acid biosynthetic process
cholesterol biosynthetic process
autophagy
lipid biosynthetic process
positive regulation of autophagy
response to activity
Wnt signaling pathway
regulation of lipid metabolic process
response to caffeine
cellular response to nutrient levels
negative regulation of TOR signaling
histone-serine phosphorylation
cellular response to glucose starvation
glucose homeostasis
regulation of circadian rhythm
negative regulation of apoptotic process
positive regulation of glycolytic process
rhythmic process
protein heterooligomerization
fatty acid homeostasis
cellular response to prostaglandin E stimulus
cellular response to organonitrogen compound
energy homeostasis
chromatin binding
protein kinase activity
protein serine/threonine kinase activity
AMP-activated protein kinase activity
protein serine/threonine/tyrosine kinase activity
protein binding
ATP binding
protein-macromolecule adaptor activity
histone serine kinase activity
metal ion binding
[hydroxymethylglutaryl-CoA reductase (NADPH)] kinase activity
[acetyl-CoA carboxylase] kinase activity
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
fatty acid biosynthetic process
cholesterol biosynthetic process
autophagy
lipid biosynthetic process
positive regulation of autophagy
response to activity
Wnt signaling pathway
regulation of lipid metabolic process
response to caffeine
cellular response to nutrient levels
negative regulation of TOR signaling
histone-serine phosphorylation
cellular response to glucose starvation
glucose homeostasis
regulation of circadian rhythm
negative regulation of apoptotic process
positive regulation of glycolytic process
rhythmic process
protein heterooligomerization
fatty acid homeostasis
cellular response to prostaglandin E stimulus
cellular response to organonitrogen compound
energy homeostasis
chromatin binding
protein kinase activity
protein serine/threonine kinase activity
AMP-activated protein kinase activity
protein serine/threonine/tyrosine kinase activity
protein binding
ATP binding
protein-macromolecule adaptor activity
histone serine kinase activity
metal ion binding
[hydroxymethylglutaryl-CoA reductase (NADPH)] kinase activity
[acetyl-CoA carboxylase] kinase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P154112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650997 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181552 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109009 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869890 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82605 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379851 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694588 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794991 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958103 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783529 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851031 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229445 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399081 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330886 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317291 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15291 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128760 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93340 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P154112 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P154112 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P154112 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P154112 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701735275 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700937421 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 98316 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 49764 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 24029 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 24543 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701735275 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700937421 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10878425 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10878417 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10878422 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10878418 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10878421 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10878423 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930558 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10878424 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10878420 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10878426 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10878419 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10878416 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10930557 | - | - |
Rat Genome 230 2.0 Array | 1369654_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369654_at | - | - |
Affymetrix HT Rat Focus Array | 1369654_at | - | - |
Affymetrix HT Rat Focus Array | 1369654_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500023690.rn.1 | TC0500003263.rn.1 | RTA TC |
Rat Expression Set 230 | 1369654_at | - | - |
Rat Expression Set 230 | 1369654_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10878416 | - | - |
Rat Genome U34 Set | rc_AA819643_at | - | - |
Rat Genome U34 Set | rc_AA819643_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21553293 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650042 | - | - |
RatRef-12 Expression BeadChip | ILMN_56898 | - | - |
