Assay Details
Gene Symbol: Cdk5rap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024488.1 | NP_077814.1 | 5 - 6 | 417 | 125 | |
GenBank mRNA | AF177476.1 | - | 5 - 6 | 417 | 125 | |
BC081793.1 | - | 4 - 5 | 339 | 7190258 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
