Assay Details
Gene Symbol: Grb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030846.2 | NP_110473.2 | 4 - 5 | 485 | 88 | |
GenBank mRNA | BC091144.1 | - | 3 - 4 | 429 | 7382476 | 88 |
D49846.1 | - | 4 - 5 | 600 | 88 | ||
FQ220840.1 | - | 4 - 5 | 551 | 88 | ||
X62853.1 | - | 3 - 4 | 335 | 88 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
Ras protein signal transduction
aging
insulin receptor signaling pathway
fibroblast growth factor receptor signaling pathway
positive regulation of signal transduction
cell differentiation
positive regulation of actin filament polymerization
receptor internalization
signal transduction in response to DNA damage
regulation of MAPK cascade
anatomical structure formation involved in morphogenesis
protein heterooligomerization
branching involved in labyrinthine layer morphogenesis
cellular response to ionizing radiation
positive regulation of reactive oxygen species metabolic process
phosphotyrosine binding
SH3/SH2 adaptor activity
epidermal growth factor receptor binding
neurotrophin TRKA receptor binding
protein binding
SH3 domain binding
protein kinase binding
protein phosphatase binding
protein domain specific binding
identical protein binding
insulin receptor substrate binding
poly(A) RNA binding
ephrin receptor binding
phosphoprotein binding
Ras protein signal transduction
aging
insulin receptor signaling pathway
fibroblast growth factor receptor signaling pathway
positive regulation of signal transduction
cell differentiation
positive regulation of actin filament polymerization
receptor internalization
signal transduction in response to DNA damage
regulation of MAPK cascade
anatomical structure formation involved in morphogenesis
protein heterooligomerization
branching involved in labyrinthine layer morphogenesis
cellular response to ionizing radiation
positive regulation of reactive oxygen species metabolic process
phosphotyrosine binding
SH3/SH2 adaptor activity
epidermal growth factor receptor binding
neurotrophin TRKA receptor binding
protein binding
SH3 domain binding
protein kinase binding
protein phosphatase binding
protein domain specific binding
identical protein binding
insulin receptor substrate binding
poly(A) RNA binding
ephrin receptor binding
phosphoprotein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P001804 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P15304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946850 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385120 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907633 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483466 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301017 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524209 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122291 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147742 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43052 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171737 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130763 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57322 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94561 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133698 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P15304 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P001804 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P15304 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P001804 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P001804 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P15304 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P15304 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P494112 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700251032 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 73364 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 46240 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 4852 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 51101 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 65033 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 75948 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700251032 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10749008 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10721231 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10749006 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10749013 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10749009 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10749016 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10721230 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10749007 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10749012 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10749015 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10749011 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10749010 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10749014 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10721229 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10749005 | - | - |
Rat Genome 230 2.0 Array | 1368386_at | - | - |
Rat Genome 230 2.0 Array | 1370617_at | - | - |
Rat Genome 230 2.0 Array | 1368385_a_at | - | - |
Rat Genome 230 2.0 Array | 1368386_PM_at | - | - |
Rat Genome 230 2.0 Array | 1370617_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368385_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1370617_at | - | - |
Affymetrix HT Rat Focus Array | 1370617_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368386_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368385_a_at | - | - |
Affymetrix HT Rat Focus Array | 1368385_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1368386_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1000037415.rn.1 | TC1000004125.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000037416.rn.1 | TC1000004125.rn.1 | RTA TC |
Rat Expression Set 230 | 1370617_at | - | - |
Rat Expression Set 230 | 1368386_at | - | - |
Rat Expression Set 230 | 1368386_PM_at | - | - |
Rat Expression Set 230 | 1370617_PM_at | - | - |
Rat Expression Set 230 | 1368385_a_at | - | - |
Rat Expression Set 230 | 1368385_PM_a_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10749005 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10721229 | - | - |
Rat Genome U34 Set | rc_AA801130_at | - | - |
Rat Genome U34 Set | rc_AI170776_s_at | - | - |
Rat Genome U34 Set | D49847_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21900160 | - | - |
ABI Rat Genome Survey Microarray | 22332231 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1352046 | - | - |
RatRef-12 Expression BeadChip | ILMN_69214 | - | - |
