Assay Details
Gene Symbol: Bhmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030850.1 | NP_110477.1 | 6 - 7 | 881 | 81 | |
GenBank mRNA | AF038870.1 | - | 6 - 7 | 881 | 81 | |
BC078811.1 | - | 6 - 7 | 868 | 7130736 | 81 | |
FQ218356.1 | - | 1 - 2 | 204 | 81 | ||
U96133.1 | - | 6 - 7 | 870 | 81 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
