Assay Details
Gene Symbol: Msn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030863.1 | NP_110490.1 | 1 - 2 | 75 | ||
GenBank mRNA | AF004811.1 | - | 1 - 2 | 75 | ||
FQ230914.1 | - | 193 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF004811.1, NM_030863.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
