Assay Details
Gene Symbol: Nfkbib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030867.2 | NP_110494.2 | 5 - 6 | 1239 | 62 | |
GenBank mRNA | AF246634.1 | - | 5 - 6 | 1062 | 62 | |
BC085729.1 | - | 5 - 6 | 1239 | 62 | ||
FQ228042.1 | - | 5 - 6 | 1035 | 62 | ||
FQ235103.1 | - | 5 - 6 | 1185 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
