Assay Details
Gene Symbol: Sigmar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030996.1 | NP_112258.1 | 1 - 2 | 190 | 65 | |
GenBank mRNA | AF004218.1 | - | 1 - 2 | 190 | 65 | |
AF067769.1 | - | 1 - 2 | 242 | 65 | ||
AF087827.1 | - | 1 - 2 | 242 | 65 | ||
BC061978.1 | - | 1 - 2 | 179 | 5623693 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
