Assay Details
Gene Symbol: P2rx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270621.1 | NP_001257550.1 | 3 - 4 | 504 | 78 | |
NM_031075.2 | NP_112337.2 | 4 - 5 | 576 | 78 | ||
GenBank mRNA | AF084975.1 | - | 3 - 4 | 349 | 78 | |
X90651.1 | - | 4 - 5 | 576 | 78 | ||
X91167.1 | - | 4 - 5 | 566 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
