Assay Details
Gene Symbol: P2rx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270621.1 | NP_001257550.1 | 4 - 5 | 591 | 64 | |
NM_031075.2 | NP_112337.2 | 5 - 6 | 663 | 64 | ||
NR_073054.1 | 6 - 7 | 685 | 64 | |||
GenBank mRNA | AF084975.1 | - | 4 - 5 | 436 | 64 | |
BC081783.1 | - | 6 - 7 | 685 | 64 | ||
X90651.1 | - | 5 - 6 | 663 | 64 | ||
X91167.1 | - | 5 - 6 | 653 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
