Assay Details
Gene Symbol: P2rx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270621.1 | NP_001257550.1 | 7 - 8 | 958 | 61 | |
NM_031075.2 | NP_112337.2 | 8 - 9 | 1030 | 61 | ||
NR_073054.1 | 9 - 10 | 1052 | 61 | |||
GenBank mRNA | AF084975.1 | - | 7 - 8 | 803 | 61 | |
BC081783.1 | - | 9 - 10 | 1052 | 7134300 | 61 | |
X90651.1 | - | 8 - 9 | 1030 | 61 | ||
X91167.1 | - | 8 - 9 | 1020 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
