Assay Details
Gene Symbol: Tgfb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031131.1 | NP_112393.1 | 4 - 5 | 1036 | 118 | |
XM_006250448.2 | XP_006250510.1 | 3 - 4 | 1862 | 118 | ||
GenBank mRNA | AF135598.1 | - | 3 - 4 | 646 | 118 | |
AF153012.2 | - | 4 - 5 | 1036 | 118 | ||
AF153013.1 | - | 3 - 4 | 952 | 118 | ||
AJ132718.1 | - | 3 - 4 | 660 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
