Assay Details
Gene Symbol: Smpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031360.2 | NP_112650.1 | 8 - 9 | 990 | 69 | |
XM_008773007.1 | XP_008771229.1 | 9 - 10 | 927 | 69 | ||
GenBank mRNA | AB047002.1 | - | 8 - 9 | 729 | 69 | |
BC091139.1 | - | 8 - 9 | 990 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
