Assay Details
Gene Symbol: Il1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031512.2 | NP_113700.2 | 1 - 2 | 43 | 62 | |
GenBank mRNA | BC091141.1 | - | 1 - 2 | 43 | 7365029 | 62 |
M98820.1 | - | 1 - 2 | 63 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC091141.1, M98820.1, NM_031512.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
