Assay Details
Gene Symbol: Ctsk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031560.2 | NP_113748.1 | 4 - 5 | 473 | 84 | |
GenBank mRNA | AF010306.1 | - | 4 - 5 | 442 | 84 | |
BC078793.1 | - | 4 - 5 | 473 | 84 | ||
FQ222438.1 | - | 4 - 5 | 417 | 84 | ||
FQ228691.1 | - | 4 - 5 | 490 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
