Assay Details
Gene Symbol: Cpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031640.1 | NP_113828.1 | 2 - 3 | 102 | 109 | |
GenBank mRNA | AF097723.1 | - | 2 - 3 | 102 | 109 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF097723.1, NM_031640.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
