Assay Details
Gene Symbol: Ndrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271091.1 | NP_001258020.1 | 14 - 15 | 1072 | 76 | |
NM_001271092.1 | NP_001258021.1 | 14 - 15 | 1072 | 76 | ||
NM_001271093.1 | NP_001258022.1 | 14 - 15 | 1019 | 76 | ||
NM_001271094.1 | NP_001258023.1 | 14 - 15 | 1019 | 76 | ||
NM_001271095.1 | NP_001258024.1 | 12 - 13 | 937 | 76 | ||
NM_031967.2 | NP_114173.2 | 12 - 13 | 937 | 76 | ||
XM_006255123.2 | XP_006255185.1 | 14 - 15 | 1014 | 76 | ||
GenBank mRNA | AF045564.1 | - | 12 - 13 | 869 | 76 | |
AY217029.1 | - | 12 - 13 | 908 | 76 | ||
AY217030.1 | - | 14 - 15 | 1019 | 76 | ||
AY217031.1 | - | 14 - 15 | 1019 | 76 | ||
AY217032.1 | - | 14 - 15 | 1067 | 76 | ||
AY217033.1 | - | 14 - 15 | 1067 | 76 | ||
AY255791.1 | - | 12 - 13 | 908 | 76 | ||
CB585874.1 | - | 8 - 9 | 494 | 76 | ||
CB713901.1 | - | 5 - 6 | 252 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
