Assay Details
Gene Symbol: Prkab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031976.1 | NP_114182.1 | 6 - 7 | 864 | 62 | |
XM_006249481.2 | XP_006249543.1 | 7 - 8 | 864 | 62 | ||
GenBank mRNA | BC062008.1 | - | 6 - 7 | 750 | 62 | |
U42411.1 | - | 6 - 7 | 864 | 62 | ||
X95577.1 | - | 7 - 8 | 834 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
