Assay Details
Gene Symbol: Src | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031977.1 | NP_114183.1 | 5 - 6 | 571 | 66 | |
XM_008762387.1 | XP_008760609.1 | 5 - 6 | 580 | 66 | ||
XM_008762388.1 | XP_008760610.1 | 5 - 6 | 617 | 66 | ||
XM_008762389.1 | XP_008760611.1 | 5 - 6 | 688 | 66 | ||
GenBank mRNA | AF130457.1 | - | 4 - 5 | 553 | 66 | |
AF157016.1 | - | 5 - 6 | 571 | 66 | ||
DQ120509.1 | - | 4 - 5 | 537 | 66 | ||
DQ120510.1 | - | 4 - 5 | 537 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
