Assay Details
Gene Symbol: Trpv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031982.1 | NP_114188.1 | 12 - 13 | 1862 | 76 | |
XM_008767882.1 | XP_008766104.1 | 11 - 12 | 1869 | 76 | ||
GenBank mRNA | AB015231.3 | - | 1113 | 76 | ||
AB040873.1 | - | 11 - 12 | 1782 | 76 | ||
AB041029.1 | - | 10 - 11 | 1602 | 76 | ||
AF029310.1 | - | 12 - 13 | 1862 | 76 | ||
AF158248.2 | - | 6 - 7 | 1195 | 76 | ||
LC008303.1 | - | 7 - 8 | 1102 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
