Assay Details
Gene Symbol: Sdcbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031986.1 | NP_114192.1 | 4 - 5 | 321 | 92 | |
GenBank mRNA | AF248548.1 | - | 4 - 5 | 321 | 92 | |
AJ292243.1 | - | 3 - 4 | 264 | 92 | ||
BC064651.1 | - | 4 - 5 | 289 | 5598152 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
