Assay Details
Gene Symbol: Hagh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033349.2 | NP_203500.2 | 2 - 3 | 281 | 72 | |
XM_006245899.1 | XP_006245961.1 | 2 - 3 | 258 | 72 | ||
XM_006245900.1 | XP_006245962.1 | 1 - 2 | 172 | 72 | ||
GenBank mRNA | BC097301.1 | - | 2 - 3 | 233 | 72 | |
CB715597.1 | - | 2 - 3 | 310 | 72 | ||
FQ229806.1 | - | 2 - 3 | 350 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
