Assay Details
Gene Symbol: Smpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053395.1 | NP_445847.1 | 3 - 4 | 313 | 63 | |
XM_006256945.2 | XP_006257007.1 | 3 - 4 | 282 | 63 | ||
GenBank mRNA | AF364071.1 | - | 3 - 4 | 313 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
