Assay Details
Gene Symbol: Pdk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053551.1 | NP_446003.1 | 7 - 8 | 876 | 74 | |
GenBank mRNA | AF034577.1 | - | 7 - 8 | 876 | 74 | |
FQ216954.1 | - | 919 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
