Assay Details
Gene Symbol: Pdlim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053650.1 | NP_446102.1 | 1 - 2 | 176 | 73 | |
XM_006253119.2 | XP_006253181.1 | 1 - 2 | 211 | 73 | ||
XM_006253120.2 | XP_006253182.1 | 1 - 2 | 209 | 73 | ||
GenBank mRNA | AF002281.1 | - | 1 - 2 | 194 | 73 | |
BC081703.1 | - | 1 - 2 | 168 | 73 | ||
FQ215471.1 | - | 1 - 2 | 191 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
