Assay Details
Gene Symbol: Pdlim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053650.1 | NP_446102.1 | 5 - 6 | 737 | 94 | |
GenBank mRNA | AF002281.1 | - | 5 - 6 | 755 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
