Assay Details
Gene Symbol: Ncf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053734.2 | NP_446186.2 | 2 - 3 | 166 | 88 | |
GenBank mRNA | AF260779.1 | - | 1 - 2 | 94 | 88 | |
AF547392.1 | - | 2 - 3 | 166 | 88 | ||
AF547393.1 | - | 2 - 3 | 166 | 88 | ||
AY029167.1 | - | 2 - 3 | 166 | 88 | ||
BC061810.1 | - | 2 - 3 | 166 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
