Assay Details
Gene Symbol: Ncf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053734.2 | NP_446186.2 | 8 - 9 | 812 | 72 | |
GenBank mRNA | AF260779.1 | - | 7 - 8 | 740 | 72 | |
AF547392.1 | - | 8 - 9 | 812 | 72 | ||
AF547393.1 | - | 8 - 9 | 812 | 72 | ||
AY029167.1 | - | 8 - 9 | 812 | 72 | ||
BC061810.1 | - | 8 - 9 | 812 | 72 |
Gene Symbol: LOC100912379 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769176.1 | XP_008767398.1 | 1 - 2 | 265 | 72 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_008769176.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
