Assay Details
Gene Symbol: Pitpnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053742.1 | NP_446194.1 | 11 - 12 | 882 | 96 | |
GenBank mRNA | D21132.1 | - | 11 - 12 | 882 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P557474 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121279 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554459 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790700 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242978 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67101 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594835 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841045 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775690 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302812 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117545 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153394 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13592 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P557474 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P557474 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P557474 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P538531 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P557474 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700544679 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701733760 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 371 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62722 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 41120 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700544679 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701733760 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10762922 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762923 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762924 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762925 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762917 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762916 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762918 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762921 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762920 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762927 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762926 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762919 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10762915 | - | - |
Rat Genome 230 2.0 Array | 1367674_at | - | - |
Rat Genome 230 2.0 Array | 1367674_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1367674_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1367674_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1200015702.rn.1 | TC1200001841.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200015704.rn.1 | TC1200001841.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200015699.rn.1 | TC1200001841.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200015700.rn.1 | TC1200001841.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200015701.rn.1 | TC1200001841.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200015703.rn.1 | TC1200001841.rn.1 | RTA TC |
Rat Expression Set 230 | 1367674_at | - | - |
Rat Expression Set 230 | 1367674_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762915 | - | - |
Rat Genome U34 Set | rc_AA998446_s_at | - | - |
Rat Genome U34 Set | D21132_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22167556 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349429 | - | - |
RatRef-12 Expression BeadChip | ILMN_61037 | - | - |
