Assay Details
Gene Symbol: Gne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053765.2 | NP_446217.1 | 5 - 6 | 1323 | 56 | |
XM_006238006.2 | XP_006238068.1 | 5 - 6 | 1141 | 56 | ||
XM_006238008.1 | XP_006238070.1 | 5 - 6 | 1107 | 56 | ||
XM_006238009.2 | XP_006238071.1 | 5 - 6 | 1484 | 56 | ||
GenBank mRNA | BC062011.1 | - | 5 - 6 | 1323 | 5621384 | 56 |
FQ224587.1 | - | 4 - 5 | 986 | 56 | ||
Y07744.1 | - | 4 - 5 | 1031 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
