Assay Details
Gene Symbol: Gps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270939.1 | NP_001257868.1 | 5 - 6 | 633 | 78 | |
NM_053969.3 | NP_446421.3 | 5 - 6 | 807 | 78 | ||
XM_006247865.1 | XP_006247927.1 | 5 - 6 | 802 | 78 | ||
XM_006247866.1 | XP_006247928.1 | 5 - 6 | 802 | 78 | ||
XM_006247867.2 | XP_006247929.1 | 5 - 6 | 662 | 78 | ||
XM_008768462.1 | XP_008766684.1 | 5 - 6 | 802 | 78 | ||
XM_008768463.1 | XP_008766685.1 | 5 - 6 | 662 | 78 | ||
GenBank mRNA | BC061746.1 | - | 5 - 6 | 756 | 78 | |
X87885.1 | - | 4 - 5 | 588 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
