Assay Details
Gene Symbol: Gps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270939.1 | NP_001257868.1 | 7 - 8 | 768 | 73 | |
NM_053969.3 | NP_446421.3 | 7 - 8 | 942 | 73 | ||
XM_006247865.1 | XP_006247927.1 | 7 - 8 | 937 | 73 | ||
XM_006247866.1 | XP_006247928.1 | 7 - 8 | 934 | 73 | ||
XM_006247867.2 | XP_006247929.1 | 7 - 8 | 797 | 73 | ||
XM_008768462.1 | XP_008766684.1 | 7 - 8 | 934 | 73 | ||
XM_008768463.1 | XP_008766685.1 | 7 - 8 | 794 | 73 | ||
GenBank mRNA | BC061746.1 | - | 7 - 8 | 891 | 73 | |
X87885.1 | - | 6 - 7 | 723 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
