Assay Details
Gene Symbol: Hadh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057186.2 | NP_476534.1 | 7 - 8 | 853 | 97 | |
GenBank mRNA | AF095449.1 | - | 7 - 8 | 853 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P13054 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643833 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281556 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181763 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400871 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966904 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701399 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916365 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153238 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21839 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21485 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P13054 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P13054 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P13054 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P13054 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P528621 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700541032 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 1329 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 33079 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15526 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 58259 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700541032 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10826832 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10826829 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10826834 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10826831 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10826833 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10826830 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10826835 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10826836 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10826828 | - | - |
Rat Genome 230 2.0 Array | 1370237_at | - | - |
Rat Genome 230 2.0 Array | 1389922_at | - | - |
Rat Genome 230 2.0 Array | 1370237_PM_at | - | - |
Rat Genome 230 2.0 Array | 1389922_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370237_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1389922_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1389922_at | - | - |
Affymetrix HT Rat Focus Array | 1370237_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0200032710.rn.1 | TC0200004703.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0200032711.rn.1 | TC0200004703.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0200032712.rn.1 | TC0200004703.rn.1 | RTA TC |
Rat Expression Set 230 | 1370237_at | - | - |
Rat Expression Set 230 | 1389922_at | - | - |
Rat Expression Set 230 | 1370237_PM_at | - | - |
Rat Expression Set 230 | 1389922_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10826828 | - | - |
Rat Genome U34 Set | rc_AA891362_at | - | - |
Rat Genome U34 Set | rc_AA848758_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21211004 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366070 | - | - |
RatRef-12 Expression BeadChip | ILMN_60853 | - | - |
