Assay Details
Gene Symbol: Il10ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057193.2 | NP_476541.2 | 2 - 3 | 244 | 75 | |
XM_006242864.2 | XP_006242926.1 | 2 - 3 | 422 | 75 | ||
GenBank mRNA | AJ305049.1 | - | 2 - 3 | 75 | ||
FQ223464.1 | - | 2 - 3 | 265 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
