Assay Details
Gene Symbol: Mylk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057209.1 | NP_476557.1 | 6 - 7 | 1190 | 70 | |
GenBank mRNA | J03886.1 | - | 6 - 7 | 1190 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
