Assay Details
Gene Symbol: Napa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080585.1 | NP_542152.1 | 1 - 2 | 143 | 96 | |
GenBank mRNA | BC063156.1 | - | 1 - 2 | 202 | 6921187 | 96 |
FQ213217.1 | - | 1 - 2 | 147 | 96 | ||
FQ216375.1 | - | 1 - 2 | 461 | 96 | ||
X89968.1 | - | 1 - 2 | 143 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
