Assay Details
Gene Symbol: Msmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080886.1 | NP_543162.1 | 2 - 3 | 346 | 103 | |
GenBank mRNA | BC063155.1 | - | 2 - 3 | 333 | 103 | |
D50559.1 | - | 2 - 3 | 346 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
