Assay Details
Gene Symbol: Sdha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130428.1 | NP_569112.1 | 14 - 15 | 1924 | 89 | |
GenBank mRNA | AB072907.1 | - | 14 - 15 | 1924 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P080678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968141 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4062 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351611 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529257 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841933 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55182 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658838 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136815 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P080678 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P080678 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P080678 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P165820 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 101487 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 40363 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68458 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10702475 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702476 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702482 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702470 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702484 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702474 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702479 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702478 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702480 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702471 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702473 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702477 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702472 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702469 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702468 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702481 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702483 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10702467 | - | - |
Rat Genome 230 2.0 Array | 1367678_PM_at | - | - |
Rat Genome 230 2.0 Array | 1367678_at | - | - |
Affymetrix HT Rat Focus Array | 1367678_at | - | - |
Affymetrix HT Rat Focus Array | 1367678_PM_at | - | - |
Rat Expression Set 230 | 1367678_at | - | - |
Rat Expression Set 230 | 1367678_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702467 | - | - |
Rat Genome U34 Set | rc_AA800250_at | - | - |
Rat Genome U34 Set | rc_AA925554_g_at | - | - |
Rat Genome U34 Set | rc_AA925554_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21623988 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357678 | - | - |
RatRef-12 Expression BeadChip | ILMN_55602 | - | - |
