Assay Details
Gene Symbol: Acaa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130433.1 | NP_569117.1 | 3 - 4 | 475 | 70 | |
GenBank mRNA | FQ216726.1 | - | 3 - 4 | 338 | 70 | |
FQ228855.1 | - | 3 - 4 | 340 | 70 | ||
X05341.1 | - | 3 - 4 | 475 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
