Assay Details
Gene Symbol: Map2k2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133283.1 | NP_579817.1 | 4 - 5 | 559 | 68 | |
XM_006240987.2 | XP_006241049.1 | 4 - 5 | 746 | 68 | ||
GenBank mRNA | BC126084.1 | - | 4 - 5 | 657 | 68 | |
D14592.1 | - | 4 - 5 | 530 | 68 | ||
L14936.1 | - | 4 - 5 | 559 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
