Assay Details
Gene Symbol: Pecr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133299.2 | NP_579833.1 | 6 - 7 | 808 | 88 | |
GenBank mRNA | AF021854.1 | - | 6 - 7 | 809 | 88 | |
AF099742.1 | - | 6 - 7 | 728 | 88 | ||
BC060546.1 | - | 6 - 7 | 802 | 6889831 | 88 | |
FQ209650.1 | - | 3 - 4 | 88 | |||
FQ210848.1 | - | 3 - 4 | 285 | 88 | ||
FQ218655.1 | - | 3 - 4 | 285 | 88 | ||
FQ218690.1 | - | 3 - 4 | 285 | 88 | ||
FQ219508.1 | - | 3 - 4 | 285 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
