Assay Details
Gene Symbol: Cd53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012523.2 | NP_036655.1 | 4 - 5 | 476 | 96 | |
GenBank mRNA | BC078900.1 | - | 4 - 5 | 427 | 96 | |
M57276.1 | - | 4 - 5 | 490 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
