Assay Details
Gene Symbol: P2rx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012721.2 | NP_036853.2 | 10 - 11 | 1156 | 61 | |
XM_008768851.1 | XP_008767073.1 | 9 - 10 | 1427 | 61 | ||
XM_008768852.1 | XP_008767074.1 | 9 - 10 | 1112 | 61 | ||
GenBank mRNA | X92070.1 | - | 10 - 11 | 1037 | 61 | |
X97376.1 | - | 10 - 11 | 1167 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
