Assay Details
Gene Symbol: Proc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012803.1 | NP_036935.1 | 4 - 5 | 309 | 59 | |
XM_006254526.2 | XP_006254588.1 | 4 - 5 | 381 | 59 | ||
XM_006254527.2 | XP_006254589.1 | 4 - 5 | 388 | 59 | ||
XM_008772018.1 | XP_008770240.1 | 4 - 5 | 451 | 59 | ||
GenBank mRNA | BC078879.1 | - | 3 - 4 | 286 | 7122006 | 59 |
X64336.1 | - | 4 - 5 | 309 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
