Assay Details
Gene Symbol: Chrna7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012832.3 | NP_036964.3 | 2 - 3 | 198 | 111 | |
XM_008759498.1 | XP_008757720.1 | 2 - 3 | 258 | 111 | ||
GenBank mRNA | AY574256.1 | - | 2 - 3 | 198 | 111 | |
AY671974.1 | - | 2 - 3 | 201 | 111 | ||
L31619.1 | - | 2 - 3 | 220 | 111 | ||
S53987.1 | - | 239 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
