Assay Details
Gene Symbol: Spp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012881.2 | NP_037013.2 | 3 - 4 | 197 | 71 | |
XM_008769996.1 | XP_008768218.1 | 3 - 4 | 270 | 71 | ||
GenBank mRNA | AB001382.1 | - | 2 - 3 | 71 | ||
BC078874.1 | - | 3 - 4 | 177 | 71 | ||
CV102664.1 | - | 3 - 4 | 197 | 71 | ||
FQ213473.1 | - | 3 - 4 | 186 | 71 | ||
FQ222192.1 | - | 3 - 4 | 213 | 71 | ||
M14656.1 | - | 3 - 4 | 170 | 71 | ||
M99252.1 | - | 3 - 4 | 206 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
