Assay Details
Gene Symbol: Vcam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012889.1 | NP_037021.1 | 1259 | 97 | ||
GenBank mRNA | BC089812.1 | - | 1277 | 7364929 | 97 | |
M84488.1 | - | 1259 | 97 | |||
X63722.1 | - | 1208 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
