Assay Details
Gene Symbol: Ctsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012939.1 | NP_037071.1 | 10 - 11 | 798 | 101 | |
XM_008766440.1 | XP_008764662.1 | 10 - 11 | 849 | 101 | ||
GenBank mRNA | BC085352.1 | - | 10 - 11 | 813 | 101 | |
FQ210730.1 | - | 10 - 11 | 808 | 101 | ||
FQ213183.1 | - | 10 - 11 | 808 | 101 | ||
FQ219177.1 | - | 10 - 11 | 808 | 101 | ||
FQ219285.1 | - | 10 - 11 | 808 | 101 | ||
FQ219548.1 | - | 10 - 11 | 808 | 101 | ||
FQ219611.1 | - | 10 - 11 | 827 | 101 | ||
FQ220588.1 | - | 7 - 8 | 577 | 101 | ||
FQ223707.1 | - | 10 - 11 | 804 | 101 | ||
FQ228686.1 | - | 10 - 11 | 868 | 101 | ||
FQ228938.1 | - | 10 - 11 | 805 | 101 | ||
M38135.1 | - | 10 - 11 | 792 | 101 | ||
Y00708.1 | - | 10 - 11 | 798 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
