Assay Details
Gene Symbol: Il1rap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167840.1 | NP_001161312.1 | 1 - 2 | 181 | 72 | |
NM_012968.1 | NP_037100.1 | 1 - 2 | 100 | 72 | ||
XM_006248504.2 | XP_006248566.1 | 1 - 2 | 169 | 72 | ||
GenBank mRNA | BC097339.1 | - | 1 - 2 | 116 | 72 | |
GU123169.1 | - | 1 - 2 | 181 | 72 | ||
U48592.1 | - | 1 - 2 | 100 | 72 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC097339.1, GU123169.1, NM_001167840.1, NM_012968.1, U48592.1, XM_006248504.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
